Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Separase Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP31719025UL
This item is not returnable.
View return policy
Description
Separase Polyclonal antibody specifically detects Separase in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
Separase | |
Polyclonal | |
Immunohistochemistry 1:2500 - 1:5000, Immunohistochemistry-Paraffin 1:2500 - 1:5000 | |
Caspase-like protein ESPL1, EC 3.4.22.49, ESP1extra spindle poles like 1, extra spindle pole bodies homolog 1 (S. cerevisiae), extra spindle poles like 1 (S. cerevisiae), Extra spindle poles-like 1 protein, FLJ46492, KIAA0165separin, Separase | |
This antibody was developed against Recombinant Protein corresponding to amino acids: TQKAAVETSFLDYGENLVQKWQVLSEVLSCSEKLVCHLGRLGSVSEAKAFCLEALKLTTKLQIPRQCALFLVLKGELELARNDIDLCQSDLQQVLFLLESCTEFGGVTQHLDSVKKVHLQKGKQQAQVPCPPQLPEEELFLRGPALELVA | |
25 μg | |
Apoptosis, Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Chromatin Research, DNA Repair, Mitotic Regulators | |
9700 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS, pH 7.2, 40% glycerol | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction