Learn More
SEPHS1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP15516720UL
 View more versions of this product
Catalog No. NBP15516720
Add to cart
Description
Description
SEPHS1 Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.
Specifications
Specifications
SEPHS1
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
Purified
SEPHS1
Synthetic peptides corresponding to SEPHS1(selenophosphate synthetase 1) The peptide sequence was selected from the C terminal of SEPHS1 (NP_036379). Peptide sequence PKYGEGHQAWIIGIVEKGNRTARIIDKPRIIEVAPQVATQNVNPTPGATS.
43 kDa
20ul
Stem Cell Markers
Primary
22929
Western Blot
Western Blot 5 ug/ml
P49903
EC 2.7.9.3, MGC4980, SELD, Selenium donor protein 1, Selenophosphate synthase 1, selenophosphate synthetase 1, selenophosphate synthetase 1 +E9a, selenophosphate synthetase 1 delta E2, selenophosphate synthetase 1 delta E8, selenophosphate synthetase 1 major, SPS1selenophosphate synthetase 1 +E9, water dikinase 1
Rabbit
IgG
Protein A purified
RUO
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Human