Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SEPHS2 Rabbit anti-Human, Biotin, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297672B
Description
SEPHS2 Polyclonal specifically detects SEPHS2 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
SEPHS2 | |
Polyclonal | |
Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
EC 2.7.9.3, Selenophosphate synthase 2, selenophosphate synthetase 2, SPS2water dikinase 2 | |
Produced in rabbits immunized with E. coli-derived Human SEPHS2 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Biotin | |
PBS | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
22928 | |
Store at 4°C in the dark. | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title