Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Septin 1 Polyclonal Antibody
Septin 1 Polyclonal Antibody
Supplier: Thermo Scientific PA534399
Description
Septin 1 Polyclonal Antibody for Western Blot, ICC/IF

Specifications
Septin 1 | |
Polyclonal | |
Unconjugated | |
SEPTIN1 | |
1-Sep; DIFF6; differentiation 6; differentiation 6 (deoxyguanosine triphosphate triphosphohydrolase); differentiation protein 6; LARP; MGC20394; Peanut-like protein 3; PNUTL3; Protein Diff6; SEP1; Sept1; septin 1; Septin1; septin-1; Serologically defined breast cancer antigen NY-BR-24 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
1731, 293507, 54204 | |
Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
P42209, Q5EB96, Q8WYJ6 | |
SEPTIN1 | |
A 17 amino acid synthetic peptide near the carboxy terminus of human SEPT1. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction