Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Septin-12 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$206.00 - $487.50

Specifications

Antigen Septin-12
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP1581720
SDP
View Documents
Novus Biologicals
NBP15811720UL
20 μL
Each for $206.00
Only null left
Add to Cart
 
NBP158117
SDP
View Documents
Novus Biologicals
NBP158117
100 μL
Each for $487.50
Only null left
Add to Cart
 
Description

Description

Septin-12 Polyclonal specifically detects Septin-12 in Human samples. It is validated for Western Blot.
Specifications

Specifications

Septin-12
Polyclonal
Rabbit
Cell Cycle and Replication
FLJ25410, septin 12, septin-12
43355
IgG
Western Blot
Unconjugated
RUO
Q8IYM1
124404
Synthetic peptides corresponding to Septin-12. The peptide sequence was selected from the middle region of 40433. Peptide sequence LQRLCRTVNVVPVIARADSLTMEEREAFRRRIQQNLRTHCIDVYPQMCFD.
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.