Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Septin-14 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA5118861

Description
Septin-14 Polyclonal antibody specifically detects Septin-14 in Human, Mouse samples. It is validated for Immunocytochemistry, Western BlotSpecifications
Septin-14 | |
Polyclonal | |
Unconjugated | |
SEPT14 | |
14-Sep; 1700016K13Rik; Sept14; septin 14; SEPTIN14; septin-14 | |
A synthesized peptide derived from human SEPT14, corresponding to a region within C-terminal amino acids. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Immunocytochemistry, Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q6ZU15, Q9DA97 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
346288, 74222 | |
-20°C | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title