Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Septin-5 Mouse anti-Human, Mouse, Rat, Clone: SP18, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP119221

 View more versions of this product

Catalog No. NBP119221


Add to cart

Description

Description

Septin-5 Monoclonal antibody specifically detects Septin-5 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications

Specifications

Septin-5
SP18
Western Blot 1:100-1:2000
Purified
43348
Immunoprecipitate of human brain
Protein G purified
RUO
Store at -20C. Avoid freeze-thaw cycles.
Monoclonal
Septin-5 (SP18)
Western Blot
Unconjugated
PBS, pH 7.2 and 50% glycerol. with 0.09% Sodium Azide
CDCREL, CDCREL1, CDCREL-1, cell division control related protein 1, Cell division control-related protein 1, HCDCREL-1, peanut-like 1, peanut-like 1 (Drosophila), Peanut-like protein 1, platelet glycoprotein Ib beta chain, PNUTL1H5, septin 5, septin-5
Mouse
IgG1
0.1 mg
Cell Cycle and Replication
Primary
5413
Human, Mouse, Rat
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only