Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Septin-5 Mouse anti-Human, Mouse, Rat, Clone: SP18, Novus Biologicals™
Mouse Monoclonal Antibody
Specifications
Antigen | Septin-5 |
---|---|
Clone | SP18 |
Dilution | Western Blot 1:100-1:2000 |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP119221
|
Novus Biologicals
NBP119221 |
0.1 mg | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
NB018386
|
Novus Biologicals
NBP1192210.025MG |
0.025 mg | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
Septin-5 Monoclonal antibody specifically detects Septin-5 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
Septin-5 | |
Western Blot 1:100-1:2000 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
CDCREL, CDCREL1, CDCREL-1, cell division control related protein 1, Cell division control-related protein 1, HCDCREL-1, peanut-like 1, peanut-like 1 (Drosophila), Peanut-like protein 1, platelet glycoprotein Ib beta chain, PNUTL1H5, septin 5, septin-5 | |
43348 | |
IgG1 | |
Protein G purified | |
Septin-5 (SP18) |
SP18 | |
Western Blot | |
Unconjugated | |
Mouse | |
Cell Cycle and Replication | |
PBS, pH 7.2 and 50% glycerol. with 0.09% Sodium Azide | |
5413 | |
Immunoprecipitate of human brain | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title