Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Septin 7 Rat anti-Horse, Human, Clone: 19A4, MilliporeSigma™
Rat Monoclonal Antibody
Supplier: MilliporeSigma MABT1557100UG
Description
Septin 7 Monoclonal antibody specifically detects Septin 7 in Horse, Human samples. It is validated for Immunoprecipitation, Western Blotting.Specifications
Septin 7 | |
Monoclonal | |
1 mg/mL | |
Purified rat monoclonal antibody IgG2c in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide. | |
A linear peptide corresponding to 19 amino acids from the C-terminal region of equine Septin 7. | |
100 μg | |
Primary | |
Stable for 1 year at 2-8°C from date of receipt. |
Immunoprecipitation, Western Blot | |
19A4 | |
Unconjugated | |
Rat | |
Protein G purified | |
RUO | |
Horse, Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction