Learn More
Septin 7 Rat anti-Horse, Human, Clone: 19A4, MilliporeSigma™
Rat Monoclonal Antibody
Manufacturer:  MilliporeSigma MABT155725UG
 View more versions of this product
Catalog No. MABT155725
Add to cart
Description
Description
Septin 7 Monoclonal antibody specifically detects Septin 7 in Horse, Human samples. It is validated for Immunoprecipitation, Western Blotting.
Specifications
Specifications
Septin 7
19A4
Unconjugated
Purified
A linear peptide corresponding to 19 amino acids from the C-terminal region of equine Septin 7.
25 μg
Stable for 1 year at 2-8°C from date of receipt.
Monoclonal
Immunoprecipitation, Western Blot
1 mg/mL
Purified rat monoclonal antibody IgG2c in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Rat
Protein G purified
RUO
Primary
Horse, Human