Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Septin 7 Rat anti-Horse, Human, Clone: 19A4, MilliporeSigma™
Rat Monoclonal Antibody
Specifications
Antigen | Septin 7 |
---|---|
Clone | 19A4 |
Concentration | 1 mg/mL |
Applications | Immunoprecipitation, Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABT155725
|
MilliporeSigma
MABT155725UG |
25 μg |
Each for $185.00
|
N/A | |||||
MABT1557100
|
MilliporeSigma
MABT1557100UG |
100 μg |
Each for $467.00
|
N/A | |||||
Description
Septin 7 Monoclonal antibody specifically detects Septin 7 in Horse, Human samples. It is validated for Immunoprecipitation, Western Blotting.Specifications
Septin 7 | |
1 mg/mL | |
Monoclonal | |
Purified | |
RUO | |
Purified rat monoclonal antibody IgG2c in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide. | |
Primary | |
Stable for 1 year at 2-8°C from date of receipt. |
19A4 | |
Immunoprecipitation, Western Blot | |
Unconjugated | |
Rat | |
Horse, Human | |
A linear peptide corresponding to 19 amino acids from the C-terminal region of equine Septin 7. | |
Protein G purified |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title