Learn More
Applied Biosystems™ SeqStudio™ Genetic Analyzer System with SmartStart and Sanger Sequencing Kit
The Applied Biosystems SeqStudio Genetic Analyzer is a 4-capillary, fluorescence-based capillary electrophoresis system designed to provide users with an integrated experience that is easy, efficient, and versatile for research in genetic analysis. This simple, fast workflow enables completion of Sanger sequencing, from sample to answer, in less than a workday.
Supplier:  Applied Biosystems™ A40032/PKG
Encompass_Preferred
Catalog No. A40032
This product is not available for sale online. Please contact Customer Service at 1-800-766-7000 or visit www.fishersci.com/quantstudio for more information and to contact a Life Science Specialist.
Description
Description
<ul><li>Simplified instrument software with interactive touch screen for greater ease of use</li><li>Minimized hands-on setup time with integrated reagents cartridge (SeqStudio Cartridge)</li><li>Application flexibility to run Sanger sequencing and fragment analysis on the same run</li><li>Connectivity: Wi-Fi or wired Ethernet connection</li><li>Web-enabled remote monitoring capability</li><li>Consumables usage tracking through radio frequency identification (RFID)</li></ul>
This bundle includes:
  • SeqStudio Genetic Analyzer Instrument
  • SmartStart SeqStudio Full Day Training
  • MicroAmp™ Optical 96-Well Reaction Plate barcode
  • Extended year warranty
  • Sanger Sequencing Kit
  • Septa for SeqStudio™ Genetic Analyzer, 96-well
Specifications
Specifications
SeqStudio System + SmartStart + Sanger Sequencing Kit
Sanger Sequencing
1 system