Learn More
NuGen Technologies IncSupplier Diversity Partner SEQUENCING RNA-SEQV2 8  DFS Item
Manufacturer:  NuGen Technologies IncSupplier Diversity Partner 710208
RNA-Seq System V2 is enabled by Ribo-SPIA technology, a rapid, simple, sensitive RNA amplification process developed by NuGEN. Using Ribo-SPIA technology with as little as 500 pg total RNA, microgram qty's of cDNA can be prepared in approx 4.5 hrs
Catalog No. 50-648-056