Learn More
Thermo Scientific™ [Ser25] Protein Kinase C (19-31)
For cellular and molecular biology applications
Supplier: Thermo Scientific™ J66676MCR
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
HPLC | |
Solid | |
[Ser25] Protein Kinase C (19-31) | |
RUO - research use only |
-30°C to -10°C | |
1559.84 | |
1 mg | |
Arg-Phe-Ala-Arg-Lys-Gly-Ser-Leu-Arg-Gln-Lys-Asn-Val |