Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Sera-Mag™ Oligo d(T) Magnetic Beads PROMO

Suited for capturing or isolating mRNA from a variety of sources. Due to their “cauliflower like” structure, Sera-Mag beads have high binding capacity. Oligo dT magnetic beads contain covalently bound oligo(dT)14.

$151.50 - $3480.00

Specifications

Concentration 1% solids (10 mg/mL)
Diameter (Metric) 1 μm
For Use With (Application) mRNA Purification
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
09-880-012LR1
View Documents
Cytiva
29648016
1 mL
Each for $151.50
Add to cart
 
09-981-145LR1
View Documents
Cytiva
29648017
5 mL
Each for $573.00
Add to cart
 
09-880-014LR1
View Documents
Cytiva
29648018
100 mL
Each for $3,480.00
Add to cart
 
Description

Description

  • Colloidally stable and will remain in suspension for extended periods of time in the absence of a magnetic field
  • High binding capacity
  • 1.0μm diameter
  • High oligo dT content for mRNA binding
Specifications

Specifications

1% solids (10 mg/mL)
1 μm
mRNA Purification
Safety and Handling

Safety and Handling

SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit