Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Sera-Mag™ Select PROMO

Delivers selective binding of DNA fragments for use with next-generation sequencing (NGS) and PCR clean-up.

$206.00 - $764.50

Specifications

For Use With (Application) Tested functionally in size selection
Storage Requirements 2 to 8°C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
09-928-106LR1
View Documents
Cytiva
29648024
5 mL
Each for $206.00
Add to cart
 
09-928-107LR1
View Documents
Cytiva
29648025
60 mL
Each for $764.50
Add to cart
 
Description

Description

Sera-Mag Select PCR clean-up and size selection reagent combines the exceptional binding characteristics of Sera-Mag Carboxyl Speedbeads with an optimized binding solution in a ready-to-use formulation. The DNA size range isolated from the process can be tailored to suit the user’s end requirements by adjusting the amount of reagent that is mixed with a fixed volume of sample.

  • Use a single reagent for PCR clean-up and size selection
  • Easily control what size DNA is retained or eliminated by adding various ratios of Sera-Mag Select to your sample, enabling NGS library and PCR fragment clean-up, as well as dual size selection of libraries
  • Uses the same protocols as existing reagents so it can be directly integrated into existing workflows with minimal disruption
  • Sera-Mag Select delivers tight distribution in size selection for high yield recovery of specific fragment sizes
  • Suitable for manual, high throughput and automated workflows
  • Sera-Mag Select should be stored in the refrigerator between 2°C to 8°C - Do not freeze - Store upright and keep bottle tightly sealed
Specifications

Specifications

Tested functionally in size selection
2 to 8°C
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit