Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytiva Sera-Mag Streptavidin-Coated Magnetic Particles
Exhibits high affinity for biotinylated target molecules. Cytiva Sera-Mag Streptavidin-Coated Magnetic Particles combine fast reaction kinetics and low non-specific binding for increased throughput and precision. Can be used as a universal base particle for coating ligands to the particle surface.
$857.00 - $1011.00
Catalog Number | Mfr. No. | Binding Property | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Binding Property | Price | Quantity & Availability | |||||
45-002-359
|
Cytiva
30152103010150 |
2500 to 3500 (Low) pmol per mg |
Pack of 25 for $857.00
|
|
|||||
45-002-360
|
Cytiva
30152104010150 |
3500 to 4500 (Med.) pmol per mg |
Pack of 25 for $1,011.00
|
|
|||||
Description
- Ideal for immunoassay and molecular biology applications, including sample preparation and assay development for genomics and proteomics
- Available with various nominal biotin binding capacities for optimizing assay development
- Low dissociation constant to tightly bound ligands
- Low non-specific binding for greater precision and accuracy
- Even streptavidin coating for reliable results
- Uniform, nominal 1μm diameter provides high surface area and excellent lot-to-lot reproducibility
The core of the particle is made by a free radical emulsion polymerization of styrene and acid monomer. A layer of magnetite is then coated onto this core.
Specifications
4°C | |
5 mL | |
1 μm |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title