Learn More
SeraCare™ ACCURUN 1™ Multi-Analyte Independent Positive Controls
Independent, multi-analyte control that helps with compliance of regulatory requirements
Manufacturer:  SeraCare Life Sciences A001-2705-P
 View more versions of this product
Catalog No. 22-554-469
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Alert:
Use the Centers for Disease Control (CDC) recommended universal precautions for handling ACCURUN 1 controls. Do not pipette by mouth; do not eat or drink in areas where specimens are being handled. Clean any spillage by immediately wiping up with 0.5% sodium hypochlorite solution. Dispose of all specimens, controls and materials used in testing as thought they contain infectious agents.
To prevent formation of potentially explosive compounds due to reactions of sodium azide and copper or lead pipes, flush waste lines with large quantities of water.
Specifications Description & Specifications
Specifications
ACCURUN 1 Series 2700
Tube
Detection of Antibodies to HIV 1 and 2, Antibodies to HTLV I and II, Antibodies to HBcAg, Antibodies to HCV, Antibodies to Cytomegalovirus, and HBsAg
CE, IVD, CE, HC
6 x 3.5mL
Human Serum or Plasma
60 Days Open Vial
Abbott Architect™ Prism™
2° to 8°C