SeraCare™ ACCURUN 1™ Multi-Analyte Independent Positive Controls

Independent, multi-analyte control that helps with compliance of regulatory requirements

Manufacturer: SeraCare Life Sciences A0012706P

 View more versions of this product

Catalog No. 22-554-470

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 

Alert

Use the Centers for Disease Control (CDC) recommended universal precautions for handling ACCURUN 1 controls. Do not pipette by mouth; do not eat or drink in areas where specimens are being handled. Clean any spillage by immediately wiping up with 0.5% sodium hypochlorite solution. Dispose of all specimens, controls and materials used in testing as thought they contain infectious agents.

To prevent formation of potentially explosive compounds due to reactions of sodium azide and copper or lead pipes, flush waste lines with large quantities of water.


    Specifications

    Product Type ACCURUN 1 Series 2700
    Certifications/Compliance CE, IVD, CE, HC
    Stability 60 Days Open Vial
    For Use With (Application) Detection of Antibodies to HIV 1 and 2, Antibodies to HTLV I and II, Antibodies to HBcAg, Antibodies to HCV, Antibodies to Cytomegalovirus, and HBsAg
    For Use With (Equipment) Abbott Architect™ Prism™
    Source Human Serum or Plasma,Human Serum or Plasma
    Quantity 3 x 1mL,3 x 1mL

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit