Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

SeraCare™ ACCURUN 1™ Series Blood Donor Multi-Marker Positive Controls

Premier multi-marker controls specifically designed for use in blood donor screening laboratories

$3234.00

Specifications

Certifications/Compliance IVD, CE, HC
Type ACCURUN™ 1 Multi-analyte Positive Control
Intended Use Detection of Antibodies to HIV 1 and 2, HTLV I and II, HBcAg, HCV, Cytomegalovirus, HBsAg
Matrix Human Serum or Plasma
Stability 60 Days Open Vial
View More Specs

Products 1
Catalog Number Mfr. No. For Use With (Equipment) Quantity Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Equipment) Quantity Price Quantity & Availability  
22-554-110
View Documents
SeraCare™
20000010
Bio-rad™ genetic system HIV-1/HIV-2 Plus O EIA 12 x 3.5mL
Pack for $3,234.00
N/A
Description

Description

  • Formulated to perform within the dynamic range of specific U.S. and internationally available test kits
  • Ready to use and contain markers commonly screened for donated blood
  • Extended open vial stability of up to 60 days
  • Contain HBsAg and antibodies to HIV-1, HIV-2, HCV, HTLV 1, HTLV II, HBc, and CMV
  • In vitro diagnostic use
  • Available in a wide variety of convenient package sizes

  • Combine LabLink xL with the ACCURUN products for an IVD exclusive infectious disease and nucleic acid testing Quality Assurance Program (QAP) from Fisher HealthCare
  • With the LabLink xL QAP program, a lab's performance can be compared to peer laboratories' performances and used to benchmark performance to group standards:
    • Track how other lab's signal to cutoff ratio (s/co) or recovered values (mean values) compare
    • Compare how a lab's variability (standard deviation) compares to a group average
    • Determine if a lab's s/co or recovered values are statistically within acceptable group limits

Specifications

Specifications

IVD, CE, HC
Detection of Antibodies to HIV 1 and 2, HTLV I and II, HBcAg, HCV, Cytomegalovirus, HBsAg
60 Days Open Vial
ACCURUN™ 1 Multi-analyte Positive Control
Human Serum or Plasma
2° to 8°C
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit