SeraCare™ ACCURUN 1™ Series Blood Donor Multi-Marker Positive Controls

Premier multi-marker controls specifically designed for use in blood donor screening laboratories

Manufacturer: SeraCare Life Sciences A0015103P

 View more versions of this product

Catalog No. 22-554-115

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    Product Type ACCURUN™ 1 multi-analyte positive control
    Certifications/Compliance IVD, CE, HC
    Stability 60 Days Open Vial
    For Use With (Application) Detection of Antibodies to HIV 1 and 2, HTLV I and II, HBcAg, HCV, Cytomegalovirus, HBsAg
    Matrix Human Serum or Plasma
    For Use With (Equipment) Abbott EIA, commander
    Storage Requirements 2° to 8°C
    Quantity 3 x 5mL,3 x 5mL

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit