Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SeraCare™ ACCURUN™ 305/306 HCV RNA Positive Control

Enhance quality control and quality assurance

Manufacturer:  SeraCare™ 20200083

 View more versions of this product

Catalog No. 22-554-140


Add to cart
This item is not returnable. View return policy

Description

Description

  • Human plasma based controls mimic patient samples
  • Independent controls meet compliance requirements of regulatory agencies and professional organizations
  • Liquid, ready-to-use: no reconstitution or special handling required
  • Extended shelf life means efficiency: allows user to monitor assay performance with same lot over an extended period of time
  • Manufactured from human serum or plasma
  • Reactive for HCV RNA type 1 and nonreactive for HBsAg and antibodies to HIV 1 and HIV 2 and HTLV
  • For in vitro diagnostic use

Specifications

Specifications

ACCURUN 305, Series 300 HCV Positive Control
IVD, CE, HC
5 x 1mL
Human Serum or Plasma
-70°C
Detect and quantitate HCV RNA
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit