Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SeraCare™ ACCURUN™ 305/306 HCV RNA Positive Control
Enhance quality control and quality assurance
$1018.00 - $1616.00
Specifications
Certifications/Compliance | IVD, CE, HC |
---|---|
Format | Vial |
Intended Use | Detect and quantitate HCV RNA |
Source | Human Serum or Plasma |
Catalog Number | Mfr. No. | Type | Quantity | Source | Content And Storage | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Type | Quantity | Source | Content And Storage | Price | Quantity & Availability | ||
22-554-516
|
SeraCare™
20200085 |
ACCURUN 305 Series 400 HCV Positive Control | 5 x 4 mL | Human Serum or Plasma | 2°C to 8°C |
Pack of 5 for $1,616.00
|
|
||
22-554-138
|
SeraCare™
20200080 |
ACCURUN 305 Series 100 HCV Positive Control | 5 x 1 mL | Human Serum or Plasma | -70°C |
Pack for $1,018.00
|
|
||
Description
- Human plasma based controls mimic patient samples
- Independent controls meet compliance requirements of regulatory agencies and professional organizations
- Liquid, ready-to-use: no reconstitution or special handling required
- Extended shelf life means efficiency: allows user to monitor assay performance with same lot over an extended period of time
- Manufactured from human serum or plasma
- Reactive for HCV RNA type 1 and nonreactive for HBsAg and antibodies to HIV 1 and HIV 2 and HTLV
- For in vitro diagnostic use
Specifications
IVD, CE, HC | |
Vial | |
Detect and quantitate HCV RNA | |
Human Serum or Plasma |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title