SeraCare™ ACCURUN™ HIV RNA Positive Control

Independent controls for HIV enhance the consistency and precision of test results, day-to-day and run-to-run

Manufacturer: SeraCare Life Sciences A3155332

 View more versions of this product

Catalog No. 22-554-519

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Specifications

    Product Type Accurun 315 Series 300
    For Use With (Application) Detect and quantitate HIV-1 RNA
    Storage Requirements 2° to 8°C
    Source Human Plasma,Human Plasma
    Certifications/Compliance IVD, CE, HC
    Format Vial
    Quantity 5 x 4mL,5 x 4mL

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit