SeraCare™ ACCURUN™ HPV Positive and Negative Controls

Independent positive and negative controls to enhance quality control and quality assurance in the diagnostic testing lab

Manufacturer: SeraCare Life Sciences A3724034E

 View more versions of this product

Catalog No. 22-554-565

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Specifications

    Product Type ACCURUN™ 372 HPV DNA Positive Control
    Certifications/Compliance RUO
    Format Vial
    For Use With (Application) Detect Human Papilloma Virus DNA in Human Cervical samples
    Quantity 10 x 0.8mL,10 x 0.8mL

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit