Learn More
SeraCare™ ACCURUN™ QC Hepatitis Positive Controls
Offer the ultimate in flexibility and convenience
Manufacturer:  SeraCare Life Sciences A1135005
 View more versions of this product
Catalog No. 22-554-650
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
ACCURUN™ 113 HBc IgM positive control
Tube
Detection of IgM Antibodies to Hepatitis B core Antigen (HBc IgM)
6 x 3.5mL
Human Serum or Plasma
60 Days Open Vial
IVD, EU, CE