Learn More
SeraCare™ ACCURUN™ QC Hepatitis Positive Controls
Offer the ultimate in flexibility and convenience
Manufacturer:  SeraCare Life Sciences A1215005
 View more versions of this product
Catalog No. 22-554-651
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
ACCURUN™ 121 Anti-HAV IgM positive control
60 Days Open Vial
IVD, EU, CE
Human Serum or Plasma
Qualitative determination of IgM Antibodies to Hepatitis A Virus (HAV IgM)
6 x 3.5mL