Learn More
SeraCare™ ACCURUN™ QC Hepatitis Positive Controls
Offer the ultimate in flexibility and convenience
Manufacturer:  SeraCare™ 20150071
 View more versions of this product
Catalog No. 22-554-167
Add to cart
Description
Description
  • Manufactured from human serum or plasma
Specifications
Specifications
ACCURUN™ 117 HBeAg Positive Control
Vial
IVD, EU, CE
1 x 5mL
Human Serum or Plasma
60 Days Open Vial
-20°C
Qualitative determination of Hepatitis B e Antigen (HBeAg)