Learn More
SeraCare™ ACCURUN™ QC Hepatitis Positive Controls
Offer the ultimate in flexibility and convenience
Manufacturer:  SeraCare Life Sciences A1105001
 View more versions of this product
Catalog No. 22-554-164
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
ACCURUN™ 110 HBsAg positive control
Vial
Qualitative detection of Hepatitis B Surface Antigen (HBsAg)
2° to 8°C
Human Serum or Plasma
60 Days Open Vial
IVD, EU, CE
1 x 5mL