SeraCare™ ACCURUN™ QC Serology Multi-Marker Negative Controls

Simplify testing by using one control vial for analytes commonly grouped in one laboratory area

Manufacturer: SeraCare Life Sciences A0011008N

 View more versions of this product

Catalog No. 22-554-126

  Check Availability
Add to Cart

    Specifications

    Product Type ACCURUN™ 1 multi-analyte negative control
    Certifications/Compliance IVD, CE, HC
    For Use With (Application) Detection of HBsAg, HIV-1 Ag and Antibodies to HIV Types 1 and 2, Antibodies to HTLV I and II, Antibodies to HBcAg, Antibodies to HCV, Antibodies to Cytomegalovirus and Antibodies to Treponema pallidum (Syphilis)
    For Use With (Equipment) Any system
    Storage Requirements 2° to 8°C
    Quantity 12 x 3.5mL,12 x 3.5mL

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit