Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SeraCare ACCURUN QC Serology Single Analyte Positive Controls
Offer the ultimate in flexibility and convenience
$145.00 - $303.00
Specifications
Certifications/Compliance | IVD, CE |
---|---|
Format | Vial |
Source | Human Serum or Plasma |
Stability | 60 Days Open Vial |
Catalog Number | Mfr. No. | Type | Quantity | Storage Requirements | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Type | Quantity | Storage Requirements | Price | Quantity & Availability | |||
22-554-175
|
SeraCare
20150080 |
ACCURUN™ 130 Lyme IgG Positive Control | 1 x 1mL | 2° to 8°C |
Pack for $145.00
|
|
22-554-176
|
SeraCare
20150081 |
ACCURUN™ 132 Lyme IgM Positive Control | 1 x 1mL | -20°C |
Pack for $229.00
|
|
22-554-179
|
SeraCare
20150084 |
ACCURUN™ 140 Rubella IgG Positive Control | 1 x 1mL | 2° to 8°C |
Pack for $244.00
|
|
22-554-181
|
SeraCare
20150086 |
ACCURUN™ 141 Rubella IgM Positive Control | 1 x 1mL | -20°C |
Pack for $210.00
|
|
22-554-182
|
SeraCare
20150087 |
ACCURUN™ 145 CMV IgG Positive Control | 1 x 1mL | 2° to 8°C |
Pack for $156.00
|
|
22-554-187
|
SeraCare
20150094 |
ACCURUN™ 156 Reagin (Syphilis) Positive Control | 3 x 1mL | -20°C |
Pack for $303.00
|
|
Description
Quality control for infectious disease testing laboratories.
- Detect and prevent errors
- Monitor assay performance
- Simplify testing procedures
- Document regulatory compliance
- Contain a single antibody or antigen and are designed to perform in the dynamic range of most manufacturers' assays
- Designed to mimic a patient sample
- Supplied ready to use, requiring no reconstitution or special handling
Specifications
IVD, CE | |
Vial | |
Human Serum or Plasma | |
60 Days Open Vial |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title