SeraCare™ ACCURUN™ QC Serology Single Analyte Positive Controls

Offer the ultimate in flexibility and convenience

Manufacturer: SeraCare Life Sciences A1655118

 View more versions of this product

Catalog No. 22-554-188

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    Product Type ACCURUN™ 165 WNV IgG/IgM positive control
    Certifications/Compliance IVD, CE
    Format Vial
    Stability 60 Days Open Vial
    For Use With (Application) Qualitative determination of IgM and IgG Antibodies to WNV
    For Use With (Equipment) Focus
    Source Human Serum or Plasma,Human Serum or Plasma
    Storage Requirements -20°C
    Quantity 3 x 0.25mL,3 x 0.25mL

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit