SeraCare™ ACCURUN™ QC Serology Single Analyte Positive Controls

Offer the ultimate in flexibility and convenience

Manufacturer: SeraCare Life Sciences A1355004

 View all options for this product

Catalog No. 22-554-177

  Check Availability
Add to Cart


    Product Type ACCURUN™ 135 Toxo IgG positive control
    Source Human Serum or Plasma
    Format Vial
    Stability 60 Days Open Vial
    For Use With (Application) Qualitative detection of IgG Antibodies to Toxoplasma gondii
    For Use With (Equipment) IMX, Axsym™, Vidas™
    Certifications/Compliance IVD, CE
    Storage Requirements 2° to 8°C
    Quantity 1 x 1mL

    Quality control for infectious disease testing laboratories.

    • Detect and prevent errors
    • Monitor assay performance
    • Simplify testing procedures
    • Document regulatory compliance
    • Contain a single antibody or antigen and are designed to perform in the dynamic range of most manufacturers' assays
    • Designed to mimic a patient sample
    • Supplied ready to use, requiring no reconstitution or special handling

  • Description & Specifications

Related Products

Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit