Learn More
SeraCare™ ACCURUN™ QC Serology Single Analyte Positive Controls
Offer the ultimate in flexibility and convenience
Manufacturer:  SeraCare Life Sciences A1365004
 View more versions of this product
Catalog No. 22-554-178
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
ACCURUN™ 136 Toxo IgM positive control
Vial
Qualitative determination of IgM Antibodies to Toxoplasma gondii
IVD, CE
1 x 1mL
Human Serum or Plasma
60 Days Open Vial
IMX, Axsym™, Vidas™
-20°C