Learn More
SeraCare™ AccuSet™ HIV p24 Antigen Mixed Titer Performance Panels Modified
For use by manufacturers and laboratories to evaluate the sensitivity and specificity of HIV antigen and HIV antigen/antibody combination assays using well-characterized specimens
Supplier: SeraCare™ 08000300
Alert:
Includes potentially infectious materials.
Description
- Includes 18 member, 1.5mL per vial panel of undiluted and naturally-occurring plasma samples collected from asymptomatic blood donors from 1997 through 2008
- Originated from HIV p24 Mixed Titer Performance Panel PRA204-1.5
- Includes Fiebig stage and test results for 13 commercially available assays for HIV p24 antigen, HIV antigen/antibody, anti-HIV, and HIV RNA
- HIV p24 antigen concentration ranges from <3 to >400 pg/mL in these samples
- Represents Fiebig stages II through VI
- Preservatives free
Specifications
HIV p24 Antigen Mixed Titer | |
18 Member |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)