Learn More
SeraCare™ Anti-Hepatitis A Virus (HAV) Mixed Titer Panel
Naturally occurring plasma specimens with reactivities ranging from negative to strongly positive for Hepatitis A IgG and/or IgM antibody
Manufacturer:  SeraCare™ 08200167
Catalog No. 22-556-291
Add to cart
This item is not returnable. View return policy
Description
Description
  • Undiluted, unpreserved plasma specimens collected between 2000 and 2010 from different individuals displaying a range of reactivities for HAV IgM and IgG
  • antibodies by several commercially available assays
  • One specimen negative for Hepatitis A IgG and IgM antibody is included as a nonreactive sample
  • For research use only: not for diagnostic procedures
Specifications
Specifications
Panel
-10°C
Evaluating Anti-Hepatitis A tests using well characterized specimens, to provide comprehensive data for comparative analysis
Set of 21 aliquots from Plasma units with reactivities ranging from negative to strongly positive for Hepatitis A IgG and/or IgM antibody, one specimen negative for Hepatitis A IgG and IgM antibody as a nonreactive sample
21 x 1.2mL
Anti-Hepatitis A Virus (HAV) Mixed Titer