Learn More
SeraCare™ HBV Genotype A Seroconversion Panels
For evaluating HBV test systems using well-characterized specimens, and to provide comprehensive data for comparative analysis
Supplier: SeraCare™ 06050040
Alert:
These materials have not been treated and should be considered biohazardous. Some panel members were found positive by tests for HBV markers, including HBsAg.
Description
- Group of close bleeds from an individual plasma donor during seroconversion
- Contains set of undiluted plasma samples from single plasma donor
- 1 vial per member, 1.5mL per vial
Cat. No. 21-123-104:
- Six member panel
- Collected over an 16-day period in 2000
- HBV DNA is positive from the first bleed
- Members PHM938-01 through -03 are challenging samples for different HBsAg assays
- PHM938-06 is a challenging HBeAg sample
- Five member panel
- Collected over an 21-day period in 2001
- HBV DNA is positive from first collection
- Other HBV markers are negative as expected in very early infection
- Members 1 through 4 are challenging for many HBsAg assays
Specifications
-20°C or -70°C | |
For evaluating HBV test system using well characterized specimens, to provide comprehensive data for comparative analysis | |
16 Days in 2000 |
HBV Seroconversion Panel | |
6 x 1.5 mL |