Learn More
SeraCare™ HBV Genotype A Seroconversion Panels
For evaluating HBV test systems using well-characterized specimens, and to provide comprehensive data for comparative analysis
$2642.00
Specifications
Type HBV Seroconversion Panel
Intended Use For evaluating HBV test system using well characterized specimens, to provide comprehensive data for comparative analysis
View More Specs
Alert:
These materials have not been treated and should be considered biohazardous. Some panel members were found positive by tests for HBV markers, including HBsAg.
Products 2
Catalog Number Mfr. No. Quantity Time Range Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Time Range Price Quantity & Availability  
21-123-105
View Documents
SeraCare™
06050041
5 x 1.5 mL 21 Days in 2001
Pack of 5 for $2,642.00
Add to cart
21-123-104
View Documents
SeraCare™
06050040
6 x 1.5 mL 16 Days in 2000 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description
Description
  • Group of close bleeds from an individual plasma donor during seroconversion
  • Contains set of undiluted plasma samples from single plasma donor
  • 1 vial per member, 1.5mL per vial
Cat. No. 21-123-104:
  • Six member panel
  • Collected over an 16-day period in 2000
  • HBV DNA is positive from the first bleed
  • Members PHM938-01 through -03 are challenging samples for different HBsAg assays
  • PHM938-06 is a challenging HBeAg sample
Cat. No. 21-123-105:
  • Five member panel
  • Collected over an 21-day period in 2001
  • HBV DNA is positive from first collection
  • Other HBV markers are negative as expected in very early infection
  • Members 1 through 4 are challenging for many HBsAg assays
Specifications
Specifications
HBV Seroconversion Panel
For evaluating HBV test system using well characterized specimens, to provide comprehensive data for comparative analysis