Seracare™ HBV Genotype A Seroconversion Panels
For evaluating HBV test systems using well-characterized specimens, and to provide comprehensive data for comparative analysis
Manufacturer: SeraCare Life Sciences PHM9391.5
 View more versions of this product
Catalog No. 21-123-105
  Check Availability
Add to Cart
Alert
These materials have not been treated and should be considered biohazardous. Some panel members were found positive by tests for HBV markers, including HBsAg.
    Specifications
    Product Type HBV seroconversion panel
    For Use With (Application) For evaluating HBV test system using well characterized specimens, to provide comprehensive data for comparative analysis
    Time Range 21 Days in 2001
    Storage Requirements -20° or -70°C
    Quantity 5 x 1.5mL,5 x 1.5mL
  • Description & Specifications
Related Products
Product Content Feedback