SeraCare™ HIV-1 Seroconversion Panels

Undiluted samples collected from a single donor during development of infection and subsequent immune response

Manufacturer: SeraCare Life Sciences PRB9681.0

 View more versions of this product

Catalog No. 22-554-549

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Specifications

    Product Type Bleed dates July/August 2007
    For Use With (Application) Evaluating HIV-1 test system using well characterized specimens, to provide comprehensive data for comparative analysis
    Storage Requirements -65° to -80°C
    Quantity 10 x 1mL,10 x 1mL

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit