Learn More
SeraCare™ QC Serology Qualification Panels
Detect testing problems before they affect laboratory operations
Specifications
Type AccuSet™ Qualification Panel
Source Human Serum or Plasma
View More Specs
Products 1
Catalog Number Mfr. No. Intended Use Quantity Storage Requirements Price Quantity & Availability  
Catalog Number Mfr. No. Intended Use Quantity Storage Requirements Price Quantity & Availability  
22-554-091
View Documents
SeraCare™
24000140
Detection of HIV-1 RNA, HCV RNA, and HBV DNA 5 x 4mL -70°C
Pack of 5 for $683.00
N/A
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
  • Convenient panels of characterized diluted samples designed to help testing laboratories:
    • Document personnel training
    • Troubleshoot assay performance
    • Evaluate proficiency
    • Document personnel training
    • Qualify incoming reagents
    • Fulfill regulatory requirements
    • Extensive array of panels helps your lab maintain the highest standards
Specifications
Specifications
AccuSet™ Qualification Panel
Human Serum or Plasma
Safety and Handling
Safety and Handling
ShelfLife : 6 Months