SeraCare™ QC Serology Qualification Panels

Detect testing problems before they affect laboratory operations

Manufacturer: SeraCare Life Sciences QHG7112.0

 View more versions of this product

Catalog No. 22-554-197

  Check Availability
Add to Cart

    Specifications

    Product Type AccuSet™ qualification panel
    For Use With (Application) Detection of Anti-HBc
    Source Human Serum or Plasma,Human Serum or Plasma
    Storage Requirements 2° to 8°C
    Quantity 6 x 2mL,6 x 2mL

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit