Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva SeraSil-Mag Silica Coated Superparamagnetic Beads PROMO

Cytiva SeraSil-Mag Silica Coated Superparamagnetic Beads PROMO deliver high purity DNA extraction in NGS applications

Specifications

Concentration 20 mg/mL in water (0.05% sodium azide)
Product Type Silica Coated, Core-Shell Structure
Quantity 60 mL
For Use With (Application) Chaotropic Salt Chemistry, Vortex Mixing, Roller Mixing, and Sonication
Storage Requirements Room Temperature
View More Specs

Products 2
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
09-928-103LR0
View Documents
Cytiva
29489259
400 nm Diameter Particle N/A N/A View substitute products »
09-928-105LR0
View Documents
Cytiva
29489260
700 nm Diameter Particle N/A N/A View substitute products »
Description

Description

  • Delivers high purity DNA extraction for highly sensitive applications where sample is scarce
  • These beads provide an optimal surface for nucleic acid binding with high performance and low background
  • High magnetization (60 emu/g) and strong binding capacity giving fast magnetic response (∼5 secs) and shorten time required for magnetic steps during isolation
  • Beads come in two sizes (submicroscale diameter 700 nm and 400 nm), offering choice based on surface requirements and high binding capacity due to the small sized particles
  • Beads are monodispersed with narrow size distribution for consistent, reproducible results
  • Silanol hydroxyl groups on the bead surface give efficient, high purity isolation of nucleic acids
  • Tested for microbial contamination
  • Good buoyancy stability (low sedimentation rate): beads remain well suspended and dispersed for over 90 mins (at 1 mg/mL concentration)
  • Specifications

    Specifications

    20 mg/mL in water (0.05% sodium azide)
    60 mL
    Room Temperature
    Silica Coated, Core-Shell Structure
    Chaotropic Salt Chemistry, Vortex Mixing, Roller Mixing, and Sonication
    Safety and Handling

    Safety and Handling

    ShelfLife : 24 months

    SDS
    Product Certifications
    Provide Content Correction

    We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Cancel Submit