Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SERBP1, Mouse anti-Human, DyLight 350, Clone: SERBP1/3493, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP279939UV
Description
SERBP1 Monoclonal antibody specifically detects SERBP1 in Human Samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin).Specifications
SERBP1 | |
SERBP1/3493 | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
Purified | |
SERBP1 | |
Recombinant fragment of human SERBP1 protein (around aa3-139) (exact sequence is proprietary) | |
Protein A or G purified | |
RUO | |
Primary | |
26135 |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 350 | |
50mM Sodium Borate | |
CGI-55, CHD3IP, chromodomain helicase DNA binding protein 3 interacting protein, DKFZP564M2423, HABP4L, PAI-1 mRNA binding protein, PAI1 RNA-binding protein 1, PAI-RBP1DKFZp564M2423, PAIRBP1FLJ90489, plasminogen activator inhibitor 1 RNA binding protein, plasminogen activator inhibitor 1 RNA-binding protein, SERPINE1 mRNA binding protein 1, SERPINE1 mRNA-binding protein 1 | |
Mouse | |
IgG2b Kappa | |
0.1 mL | |
Store at 4°C in the dark. | |
Monoclonal | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title