Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SERF2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA567997
Description
Predicted to react with Mouse samples.
SERF2 (small EDRK-rich factor 2) is the human ortholog of the C/ elegans MOAG-4 gene. Serf2 (small EDRK-rich factor 2) is the human ortholog of the C. elegans MOAG-4 gene. MOAG-4 was shown to be a positive regulator of protein aggregates. Inactivation of MOAG-4 leads to a rescue from proteotoxcity, and could play a role in human diseases such as Alzheimer's disease and Parkinson's disease. In addition, SERF2 can induce huntingtin aggregation in SK-N-SH neuroblastoma cells.
Specifications
SERF2 | |
Polyclonal | |
Unconjugated | |
SERF2 | |
4F5REL; 4F5rel (4F5 related); C80083; FAM2C; Gastric cancer-related protein VRG107; H4F5REL; HsT17089; m4F5rel; Msmac1l; Protein 4F5-related; SERF2; Small EDRK-rich factor 2 | |
Rabbit | |
affinity chromatography | |
RUO | |
10169, 378702 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P84101, P84102 | |
SERF2 | |
Synthesized peptide derived from human SERF2 (aa 1-30). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction