Learn More
Fisherbrand™ Serialized Class A Cylinders with Certificate of Traceability PROMO
Calibrated “To Contain”
Specifications
Material Borosilicate Glass
Autoclavable Yes
View More Specs
Products 8
Catalog Number Capacity (Metric) Graduations Limit of Error Price Quantity & Availability  
Catalog Number Capacity (Metric) Graduations Limit of Error Price Quantity & Availability  
08559APM View Documents 10 mL 0.1 mL ±0.10 mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
08559BPM View Documents 25 mL 0.2 mL ±0.17 mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
08559CPM View Documents 50 mL 1 mL ±0.25 mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
08559DPM View Documents 100 mL 1 mL ±0.50 mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
08559EPM View Documents 250 mL 2 mL ±1.0 mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
08559FPM View Documents 500 mL 5 mL ±2.0 mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
08559GPM View Documents 1000 mL 10 mL ±3.0 mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
08559HPM View Documents 2000 mL 20 mL ±6.0 mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Serialized Class A
  • With traceability
  • Borosilicate glass
  • White scale
Specifications
Specifications
Borosilicate Glass
Yes