Learn More
Fisherbrand™ Serialized Class A Cylinders with Certificate of Traceability PROMO
Calibrated “To Deliver”
Specifications
Material Borosilicate Glass
Class A
Includes Traceable NIST certificate
Scale White scale graduations
Sterility Sterile
View More Specs
Products 8
Catalog Number Capacity (Metric) Graduations Tolerance (Metric) Subdivision Price Quantity & Availability  
Catalog Number Capacity (Metric) Graduations Tolerance (Metric) Subdivision Price Quantity & Availability  
08557APM View Documents 10 mL 0.1 mL ±0.1mL 0.1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
08557BPM View Documents 25 mL 0.2 mL ±0.17mL 0.2mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
08557CPM View Documents 50 mL 1 mL ±0.25mL 1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
08557DPM View Documents 100 mL 1 mL ±0.5mL 1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
08557EPM View Documents 250 mL 2 mL ±1mL 2mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
08557FPM View Documents 500 mL 5 mL ±2mL 5mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
08557HPM View Documents 2000 mL 20 mL ±6mL 20mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
0973030PM View Documents 1000 mL 10 mL ±3mL 10mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
Calibrated “To Deliver”
  • Serialized
  • White scale
  • Molded hexagonal base
Specifications
Specifications
Borosilicate Glass
Traceable NIST certificate
Sterile
A
White scale graduations
Yes