Learn More
Binder™ Series KB Refrigerated Incubators
Binder™ Series KB Refrigerated Incubators offer a wide range of capabilities and deliver reproducible test results.
$5448.00 - $15441.00
Specifications
Description Refrigerated Incubator
Electrical Requirements 120 V, 60 Hz
Includes 2 stainless steel racks
No. of Doors 1
View More Specs
Products 6
Catalog Number Mfr. No. Model Price Quantity & Availability  
Catalog Number Mfr. No. Model Price Quantity & Availability  
08-111-000 Accessories View Documents Binder™
90200113
KB 23 Each for $5,448.00
Add to cart
08-111-001 Accessories View Documents Binder™
90200302
KB 53 Each for $5,916.00
Add to cart
08-111-002 Accessories View Documents Binder™
90200398
KB 115 Each for $7,071.00
Add to cart
08-111-003 Accessories View Documents Binder™
90200304
KB 240 Each for $10,150.00
Add to cart
08-111-004 Accessories View Documents Binder™
90200305
KB 400 Each for $11,585.00
Add to cart
08-111-005 Accessories View Documents Binder™
90200306
KB720UL-240V Each for $15,441.00
Add to cart
Description
Description
  • APT.line™ preheating chamber technology
  • Refrigeration with compressor refrigerated unit
  • Adjustable fan speed
  • Controller with time-segment and real-time programming
  • Elapsed time indicator
  • Inner door made of tempered safety glass
  • 2 stainless steel racks
  • Stackable
  • Class 3.1 independent temperature safety device (DIN 12880) with visual and audible temperature alarm
  • Computer interface: Ethernet
Specifications
Specifications
Refrigerated Incubator
120 V, 60 Hz
2 stainless steel racks
1