Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SERINC4 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549296

Catalog No. PA549296

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Incorporates a polar amino acid serine into membranes and facilitates the synthesis of two serine-derived lipids, phosphatidylserine and sphingolipids.
Specifications

Specifications

SERINC4
Polyclonal
PBS with 0.09% sodium azide
A6NH21
Serine incorporator 4, SERINC4
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
619189
Western Blot
Unconjugated
SERINC4
Liquid
SERINC4
KLH conjugated synthetic peptide between 489-518 amino acids from the C-terminal region of human SERINC4.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit