Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Serine Dehydratase Antibody (OTI3D3), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274074MFV450
Description
Serine Dehydratase Monoclonal antibody specifically detects Serine Dehydratase in Human samples. It is validated for Flow CytometrySpecifications
Serine Dehydratase | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
10993 | |
Store at 4C in the dark. | |
IgG1 |
Flow Cytometry | |
OTI3D3 | |
Flow Cytometry | |
EC 4.3.1.17, EC 4.3.1.19, L-serine ammonia-lyase, L-serine deaminase, L-serine dehydratase/L-threonine deaminase, L-threonine dehydratase, SDHL-serine dehydratase, serine dehydratase, TDH | |
Full length human recombinant protein of human SDS(NP_006834) produced in HEK293T cell. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction